Gluconic acid, sodium salt, 98%
Gluconic acid, sodium salt, 98%
Gluconic acid, sodium salt, 98%
Gluconic acid, sodium salt, 98%
Thermo Scientific Chemicals
Gluconic acid, sodium salt, 98%
CAS: 527-07-1 | C6H11NaO7 | 218.137 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA181390010
Price (MYR)
308.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
308.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGluconic acid, sodium salt
CAS527-07-1
Melting Point206.0°C to 209.0°C
ColorWhite to Beige
pH6.5 to 7.5 (10% soln.)