N,N-Diethylformamide, 99%
N,N-Diethylformamide, 99%
N,N-Diethylformamide, 99%
N,N-Diethylformamide, 99%
Thermo Scientific Chemicals
N,N-Diethylformamide, 99%
CAS: 617-84-5 | C5H11NO | 101.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA181375000
Price (MYR)
2,350.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,350.00
EA
Add to cart
Specifications
Chemical Name or MaterialN, N-Diethylformamide
Name Note99%
CAS617-84-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning