n-Butyllithium, 1.6M solution in hexanes, AcroSeal™
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™
Thermo Scientific Chemicals
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™
CAS: 109-72-8 | C4H9Li | 64.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA181271000100 mLAcroSeal™ Glass bottle
Catalog number FSA181271000
Price (MYR)
618.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
618.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Butyllithium
Name Note1.6M solution in hexanes
CAS109-72-8
CAS Min %14.75
Health Hazard 1Danger