3,5-Dimethylpiperidine, 96%, mixture of cis and trans
3,5-Dimethylpiperidine, 96%, mixture of cis and trans
3,5-Dimethylpiperidine, 96%, mixture of cis and trans
3,5-Dimethylpiperidine, 96%, mixture of cis and trans
Thermo Scientific Chemicals
3,5-Dimethylpiperidine, 96%, mixture of cis and trans
CAS: 35794-11-7 | C7H16N | 114.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA181231000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 5-Dimethylpiperidine
Name Note96%
CAS35794-11-7
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger