2-Furaldehyde, 99%
2-Furaldehyde, 99%
2-Furaldehyde, 99%
Thermo Scientific Chemicals
2-Furaldehyde, 99%
CAS: 98-01-1 | C5H4O2 | 96.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA181102500
Price (MYR)
290.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
290.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Furaldehyde
Acidity0.02 eq/L max.
CAS98-01-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger