N,N-Dimethylacetamide, 99+%, extra pure
N,N-Dimethylacetamide, 99+%, extra pure
N,N-Dimethylacetamide, 99+%, extra pure
N,N-Dimethylacetamide, 99+%, extra pure
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number FSA181070050
Price (MYR)
1,026.00
LT
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
1,026.00
LT
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylacetamide
Melting Point-20.0°C
CAS127-19-5
CAS Min %99.0
Health Hazard 1Signal Word: WARNING!