1,10-Decanediol, 99%
1,10-Decanediol, 99%
1,10-Decanediol, 99%
1,10-Decanediol, 99%
Thermo Scientific Chemicals
1,10-Decanediol, 99%
CAS: 112-47-0 | C10H22O2 | 174.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA181041000
Price (MYR)
411.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
411.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 10-Decanediol
CAS112-47-0
CAS Min %98.5
CAS Max %100.0
Melting Point71°C to 75°C