Cyanuric acid, 98%
Cyanuric acid, 98%
Cyanuric acid, 98%
Thermo Scientific Chemicals
Cyanuric acid, 98%
CAS: 108-80-5 | C3H3N3O3 | 129.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA181022500
Price (MYR)
497.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
497.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCyanuric acid
Name Note98%
CAS108-80-5
Health Hazard 1HYGROSCOPIC
ColorWhite to Beige