Cumene, 99%, pure
Cumene, 99%, pure
Cumene, 99%, pure
Thermo Scientific Chemicals
Cumene, 99%, pure
CAS: 98-82-8 | C9H12 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA181010025
Price (MYR)
861.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
861.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCumene
Name Note99%
CAS98-82-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.