2-Chlorophenol, 98+%
2-Chlorophenol, 98+%
2-Chlorophenol, 98+%
2-Chlorophenol, 98+%
Thermo Scientific Chemicals
2-Chlorophenol, 98+%
CAS: 95-57-8 | C6H5ClO | 128.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA180991000
Price (MYR)
227.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
227.00
EA
Add to cart
Specifications
Chemical Name or Material2-Chlorophenol
Name Note98+%
CAS95-57-8
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning