1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
Thermo Scientific Chemicals
1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
CAS: 90-13-1 | C10H7Cl | 162.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA180971000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Chloronaphthalene
CAS90-13-1
Health Hazard 1GHS Signal Word: Warning
CAS91-58-7
Melting Point-2.5°C