Naphthalene, 99%
Naphthalene, 99%
Naphthalene, 99%
Thermo Scientific Chemicals
Naphthalene, 99%
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA180902500
Price (MYR)
231.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
231.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNaphthalene
CAS91-20-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0