Benzyl chloride, 99.5+%, extra pure, stabilized
Benzyl chloride, 99.5+%, extra pure, stabilized
Benzyl chloride, 99.5+%, extra pure, stabilized
Benzyl chloride, 99.5+%, extra pure, stabilized
Thermo Scientific Chemicals
Benzyl chloride, 99.5+%, extra pure, stabilized
CAS: 100-44-7 | C7H7Cl | 126.583 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA180850010
Price (MYR)
366.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
366.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzyl chloride
Name Note99.5+%
CAS100-44-7
CAS Min %99.5
Health Hazard 1Danger
View more
Alkylation reagent
RUO – Research Use Only