2-Methoxyethanol, 99+%, extra pure
2-Methoxyethanol, 99+%, extra pure
2-Methoxyethanol, 99+%, extra pure
Thermo Scientific Chemicals
2-Methoxyethanol, 99+%, extra pure
2-Methoxyethanol, 99+%, C3H8O2, CAS Number-109-86-4, 2-methoxy-1-ethanol, 3-oxa-1-butanol, dowanol em, egme, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methoxyethanol, methyl cellosolve, methyl oxitol, monomethyl glycol, 2.5L, 99% min. (GC), 01, 467, 124.0 deg.C, 100.0, 99.0, CHEBI:46790 | CAS: 109-86-4 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA180790025
Price (MYR)
644.00
Bottle
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
644.00
Bottle
Specifications
Chemical Name or Material2-Methoxyethanol
Name Note99+%
CAS109-86-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger