Phenol, 99+%, for biochemistry, loose crystals
Phenol, 99+%, for biochemistry, loose crystals
Phenol, 99+%, for biochemistry, loose crystals
Phenol, 99+%, for biochemistry, loose crystals
Thermo Scientific Chemicals
Phenol, 99+%, for biochemistry, loose crystals
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA180785000
Price (MYR)
928.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
928.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenol
Name Note99+%
CAS108-95-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated