1-Tetradecanol, 99%
1-Tetradecanol, 99%
1-Tetradecanol, 99%
Thermo Scientific Chemicals
1-Tetradecanol, 99%
CAS: 112-72-1 | C14H30O | 214.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA180731000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Tetradecanol
CAS112-72-1
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0