Isopropyl ether, 99+%, extra pure, stabilized with BHT
Isopropyl ether, 99+%, extra pure, stabilized with BHT
Isopropyl ether, 99+%, extra pure, stabilized with BHT
Isopropyl ether, 99+%, extra pure, stabilized with BHT
Thermo Scientific Chemicals
Isopropyl ether, 99+%, extra pure, stabilized with BHT
CAS: 108-20-3 | C6H14O | 102.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal drum
Catalog number FSA180680250
Price (MYR)
3,924.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (MYR)
3,924.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropyl ether
Name NoteStabilized with BHT
Acidity0.005% max.
CAS108-20-3
Health Hazard 1Danger