Dimethyl terephthalate, 99%
Dimethyl terephthalate, 99%
Dimethyl terephthalate, 99%
Dimethyl terephthalate, 99%
Thermo Scientific Chemicals
Dimethyl terephthalate, 99%
CAS: 120-61-6 | C10H10O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA180585000
Price (MYR)
402.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
402.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethyl terephthalate
Name Note99%
Acidity0.03mg KOH/g max.
CAS120-61-6
Melting Point140°C to 143°C