p-Dithiane-2,5-diol, 96%
p-Dithiane-2,5-diol, 96%
p-Dithiane-2,5-diol, 96%
p-Dithiane-2,5-diol, 96%
Thermo Scientific Chemicals
p-Dithiane-2,5-diol, 96%
CAS: 40018-26-6 | C4H8O2S2 | 152.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA179932500
Price (MYR)
1,278.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,278.00
BOT
Add to cart
Specifications
Chemical Name or Materialp-Dithiane-2,5-diol
CAS40018-26-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.
Health Hazard 3GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective