(+)-Limonene, 96%, unstabilized
(+)-Limonene, 96%, unstabilized
(+)-Limonene, 96%, unstabilized
(+)-Limonene, 96%, unstabilized
Thermo Scientific Chemicals
(+)-Limonene, 96%, unstabilized
CAS: 5989-27-5 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FSA179395000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material(+)-Limonene
CAS5989-27-5
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0