1-Methylisatin, 98%
1-Methylisatin, 98%
1-Methylisatin, 98%
Thermo Scientific Chemicals
1-Methylisatin, 98%
CAS: 2058-74-4 | C9H7NO2 | 161.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA179350100
Price (MYR)
1,164.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,164.00
EA
Add to cart
Specifications
Chemical Name or Material1-Methylisatin
CAS2058-74-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0