Zein, purified
Zein, purified
Zein, purified
Thermo Scientific Chemicals
Zein, purified
CAS: 9010-66-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA179315000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialZein
Name NotePurified
CAS9010-66-6
EINECS Number232-722-9
Ignition Residue2% max. (800°C)