Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
Thermo Scientific Chemicals
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
CAS: 25067-59-8 | (C14H11N)n | 193.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number FSA179030100
Price (MYR)
1,168.00
BOT
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
1,168.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(N-vinylcarbazole)
ColorWhite
Name NoteSecondary standard
CAS25067-59-8
Physical FormCrystalline Powder