Poly(methyl methacrylate), beads, average M.W. 35,000
Poly(methyl methacrylate), beads, average M.W. 35,000
Poly(methyl methacrylate), beads, average M.W. 35,000
Poly(methyl methacrylate), beads, average M.W. 35,000
Thermo Scientific Chemicals
Poly(methyl methacrylate), beads, average M.W. 35,000
CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA178765000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(methyl methacrylate)
ColorWhite
Name Notenominal M.W. 38000
CAS9011-14-7
Physical FormBeads