Poly(ethylene oxide), M.W. 100,000 to 200,000
Poly(ethylene oxide), M.W. 100,000 to 200,000
Poly(ethylene oxide), M.W. 100,000 to 200,000
Poly(ethylene oxide), M.W. 100,000 to 200,000
Thermo Scientific Chemicals
Poly(ethylene oxide), M.W. 100,000 to 200,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
500 g
Packaging:
Glass bottle
Catalog number FSA178582500
Price (MYR)
822.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
822.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(ethylene oxide)
ColorWhite to Yellow
Name NoteApproximate M.W. 100.000
CAS25322-68-3
Physical FormPowder