Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched
Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched
Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched
Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched
Thermo Scientific Chemicals
Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched
CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA178572500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethyleneimine
Name Note50 wt. % Aqueous Solution, Average M.W. 50-60000
CAS9002-98-6
Physical FormViscous Liquid
Boiling Point100.0°C