Polycaprolactone, M.W. approx. 80,000
Polycaprolactone, M.W. approx. 80,000
Thermo Scientific Chemicals
Polycaprolactone, M.W. approx. 80,000
CAS: 24980-41-4 | (C6H10O2)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA178305000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolycaprolactone
CAS24980-41-4
Melting Point60.0°C
Molecular Formula(C6H10O2)n
Linear Formula[-O(CH2)5CO-]n