Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
Thermo Scientific Chemicals
Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
CAS: 9003-05-8 | (C3H5NO)n | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
250 g
Packaging:
Glass bottle
Catalog number FSA178045000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(acrylamide)
ColorWhite to Yellow
CAS9003-05-8
Physical FormGranules
RTECS NumberAS3700000