Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Thermo Scientific Chemicals
Cellulose acetate, average M.W. 100,000
CAS: 9004-35-7 | (C6H7O5)nC2H3O
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Packaging:
Plastic bottle
Catalog number FSA177785000
Price (MYR)
1,092.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,092.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCellulose acetate
ColorWhite
CAS9004-35-7
Physical FormPowder
TSCATSCA