2-Methoxyphenylacetic acid, 99%
2-Methoxyphenylacetic acid, 99%
2-Methoxyphenylacetic acid, 99%
2-Methoxyphenylacetic acid, 99%
Thermo Scientific Chemicals
2-Methoxyphenylacetic acid, 99%
CAS: 93-25-4 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA177601000
Price (MYR)
1,039.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,039.00
EA
Add to cart
Specifications
Chemical Name or Material2-Methoxyphenylacetic acid
CAS93-25-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0