Diphenyl diselenide, 98+%
Diphenyl diselenide, 98+%
Diphenyl diselenide, 98+%
Diphenyl diselenide, 98+%
Thermo Scientific Chemicals
Diphenyl diselenide, 98+%
CAS: 1666-13-3 | C12H10Se2 | 312.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA177570050
Price (MYR)
412.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
412.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiphenyl diselenide
CAS1666-13-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and