Carbon disulfide, 99.9%, extra pure, low in benzene content
Carbon disulfide, 99.9%, extra pure, low in benzene content
Carbon disulfide, 99.9%, extra pure, low in benzene content
Carbon disulfide, 99.9%, extra pure, low in benzene content
Thermo Scientific Chemicals
Carbon disulfide, 99.9%, extra pure, low in benzene content
CAS: 75-15-0 | CS2 | 76.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA177280025
Price (MYR)
1,812.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,812.00
EA
Add to cart
Specifications
Chemical Name or MaterialCarbon disulfide
Melting Point-111.0°C
Name NoteLow in Benzene Content
CAS75-15-0
Health Hazard 1Danger