3-Bromoaniline, 98%
3-Bromoaniline, 98%
3-Bromoaniline, 98%
Thermo Scientific Chemicals
3-Bromoaniline, 98%
CAS: 591-19-5 | C6H6BrN | 172.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA177210250
Price (MYR)
671.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
671.00
EA
Add to cart
Specifications
Chemical Name or Material3-Bromoaniline
CAS591-19-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0