Potassium hydrogen phthalate, 99.99%, acidimetric standard
Potassium hydrogen phthalate, 99.99%, acidimetric standard
Potassium hydrogen phthalate, 99.99%, acidimetric standard
Potassium hydrogen phthalate, 99.99%, acidimetric standard
Thermo Scientific Chemicals
Potassium hydrogen phthalate, 99.99%, acidimetric standard
CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA177125000
Price (MYR)
717.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
717.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPotassium Hydrogen Phthalate
CAS877-24-7
Health Hazard 1Signal Word: CAUTION!
Health Hazard 3Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated.
NFPA
Health:
Melting Point295.0°C to 300.0°C
View more
For ultra trace analysis
RUO – Research Use Only