Methanol, 99.9%, for analysis
Methanol, 99.9%, for analysis
Methanol, 99.9%, for analysis
Methanol, 99.9%, for analysis
Thermo Scientific Chemicals
Methanol, 99.9%, for analysis
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA176840025
Price (MYR)
412.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
412.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
CAS67-56-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
View more
For analysis
RUO – Research Use Only