Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Thermo Scientific Chemicals
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA176630010
Price (MYR)
513.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
513.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetrahydrofuran
Melting Point-108.4°C
CAS109-99-9
Health Hazard 1Danger
Health Hazard 2GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.