Ethylene glycol diacetate, 99%
Ethylene glycol diacetate, 99%
Ethylene glycol diacetate, 99%
Ethylene glycol diacetate, 99%
Thermo Scientific Chemicals
Ethylene glycol diacetate, 99%
CAS: 111-55-7 | C6H10O4 | 146.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA176225000
Price (MYR)
281.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
281.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthylene glycol diacetate
CAS111-55-7
Melting Point-41°C
Density1.1200g/mL
EINECS Number203-881-1