Pyridine-d5, for NMR, 100.0 atom % D
Pyridine-d5, for NMR, 100.0 atom % D
Pyridine-d5, for NMR, 100.0 atom % D
Pyridine-d5, for NMR, 100.0 atom % D
Thermo Scientific Chemicals
Pyridine-d5, for NMR, 100.0 atom % D
CAS: 7291-22-7 | C5H5N | 84.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 mL
10 mL
Packaging:
Glass Bottle
Catalog number FSA176170250
Price (MYR)
5,457.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
5,457.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyridine-d5
Density1.0500g/mL
Deuterium (D)99.95% min.
Flash Point20°C
PackagingGlass Bottle
View more
For NMR
RUO – Research Use Only