Dichloromethane-d2, for NMR, 99.5 atom % D
Dichloromethane-d2, for NMR, 99.5 atom % D
Dichloromethane-d2, for NMR, 99.5 atom % D
Dichloromethane-d2, for NMR, 99.5 atom % D
Thermo Scientific Chemicals
Dichloromethane-d2, for NMR, 99.5 atom % D
CAS: 1665-00-5 | CH2Cl2 | 86.939 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
10 mL
Packaging:
Glass Bottle
Catalog number FSA176110250
Price (MYR)
2,391.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,391.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDichloromethane-d2
Density1.3600g/mL
Deuterium (D)99.60% min.
PackagingGlass Bottle
Physical FormLiquid
View more
For NMR
RUO – Research Use Only