2,5-Dimethylfuran, 99%
2,5-Dimethylfuran, 99%
2,5-Dimethylfuran, 99%
2,5-Dimethylfuran, 99%
Thermo Scientific Chemicals
2,5-Dimethylfuran, 99%
CAS: 625-86-5 | C6H8O | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA176031000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 5-Dimethylfuran
Name Note99%
CAS625-86-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger