4-Acetylbenzoic acid, 98%
4-Acetylbenzoic acid, 98%
4-Acetylbenzoic acid, 98%
4-Acetylbenzoic acid, 98%
Thermo Scientific Chemicals
4-Acetylbenzoic acid, 98%
CAS: 586-89-0 | C9H8O3 | 164.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA17586010010 gGlass bottle
Catalog number FSA175860100
Price (MYR)
963.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
963.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Acetylbenzoic acid
Name Note98%
CAS586-89-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning