3-Quinolinecarboxylic acid, 98%
3-Quinolinecarboxylic acid, 98%
3-Quinolinecarboxylic acid, 98%
3-Quinolinecarboxylic acid, 98%
Thermo Scientific Chemicals
3-Quinolinecarboxylic acid, 98%
CAS: 6480-68-8 | C10H7NO2 | 173.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA175700050
Price (MYR)
713.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
713.00
EA
Add to cart
Specifications
Chemical Name or Material3-Quinolinecarboxylic acid
CAS6480-68-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0