Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D
Thermo Scientific Chemicals
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D
CAS: 7698-05-7 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA17542050050 gGlass Bottle
Catalog number FSA175420500
Price (MYR)
1,201.00
G
Add to cart
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,201.00
G
Add to cart
Specifications
Chemical Name or MaterialDeuterium chloride
Density1.2000g/mL
Deuterium (D)99.0% min.
PackagingGlass Bottle
Physical FormLiquid
View more
For NMR
RUO – Research Use Only