Deuterium chloride, for NMR, 20 wt. % solution in D2O, 100.0 atom % D
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 100.0 atom % D
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 100.0 atom % D
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 100.0 atom % D
Thermo Scientific Chemicals
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 100.0 atom % D
CAS: 7698-05-7 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1753400505 gGlass Bottle
Catalog number FSA175340050
Price (MYR)
487.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
487.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDeuterium chloride
Density1.2000g/mL
Deuterium (D)99.95% min.
PackagingGlass Bottle
Physical FormLiquid
View more
For NMR
RUO – Research Use Only