L(+)-Asparagine monohydrate, 99%
L(+)-Asparagine monohydrate, 99%
L(+)-Asparagine monohydrate, 99%
L(+)-Asparagine monohydrate, 99%
Thermo Scientific Chemicals
L(+)-Asparagine monohydrate, 99%
L(+)-Asparagine monohydrate, 99%, C4H10N2O4, CAS Number-5794-13-8, asparagine monohydrate, l +-asparagine monohydrate, asparagine, asparagine nf, l-asparagine hydrate, h-asn-oh.h2o, unii-2pd79vf521, l-asparagine monohydrate, l-asparagine, monohydrate, s-2,4-diamino-4-oxobutanoic acid hydrate, 250g | CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Glass Bottle
Catalog number FSA175272500
Price (MYR)
993.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
993.00
BOT
Add to cart
Specifications
Chemical Name or MaterialL(+)-Asparagine monohydrate
Melting Point233.0°C to 235.0°C
Name Note99%
Ammonium1000ppm max.
CAS5794-13-8