Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™
CAS: 14044-65-6 | C4H11BO | 85.94 g/mol
Have Questions?
Change view
Quantity:
800 mL
100 mL
Catalog number FSA175088000
Price (MYR)
2,134.00
BOT
Add to cart
Quantity:
800 mL
Request bulk or custom format
Price (MYR)
2,134.00
BOT
Add to cart
Chemical Identifiers
CAS14044-65-6, 109-99-9
Specifications
Chemical Name or MaterialBorane-tetrahydrofuran complex
ColorColorless
Name Note1M solution in tetrahydrofuran, stabilized
CAS14044-65-6
Health Hazard 1Danger