Trifluoromethanesulfonic anhydride, 98+%
Trifluoromethanesulfonic anhydride, 98+%
Trifluoromethanesulfonic anhydride, 98+%
Trifluoromethanesulfonic anhydride, 98+%
Thermo Scientific Chemicals
Trifluoromethanesulfonic anhydride, 98+%
Trifluoromethanesulfonic anhydride, 98+%, Quantity: 250mL, Packaging: Glass bottle, Boiling Point: 80.0 deg.C, Colorless to Brown, Molecular Weight: 282.13, Percent Purity: 98+%, Assay Percent Range: 98+%, Beilstein: 03, IV, 35, CAS: 358-23-6, ChEBI: CHEBI:48509, Density: 1.7000g/mL | CAS: 358-23-6 | C2F6O5S2 | 282.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 mL
50 mL
Packaging:
Glass bottle
Catalog number FSA175062500
Price (MYR)
6,263.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,263.00
EA
Add to cart
Specifications
Chemical Name or MaterialTrifluoromethanesulfonic anhydride
CAS358-23-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
May be corrosive to metals.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
View more
Alkylation reagent, Coupling reagent
RUO – Research Use Only