Methyl sulfoxide-d6, for NMR, 99+ atom % D
Methyl sulfoxide-d6, for NMR, 99+ atom % D
Methyl sulfoxide-d6, for NMR, 99+ atom % D
Methyl sulfoxide-d6, for NMR, 99+ atom % D
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, 99+ atom % D
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
Packaging:
Glass Bottle
Catalog number FSA174892500
Price (MYR)
5,555.00
ML
Add to cart
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
5,555.00
ML
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide-d6
Density1.1900g/mL
Deuterium (D)99.0% min.
Flash Point88°C
PackagingGlass Bottle
View more
For NMR
RUO – Research Use Only