Isopropanol-d8, for NMR, 99+ atom % D
Isopropanol-d8, for NMR, 99+ atom % D
Isopropanol-d8, for NMR, 99+ atom % D
Isopropanol-d8, for NMR, 99+ atom % D
Thermo Scientific Chemicals
Isopropanol-d8, for NMR, 99+ atom % D
CAS: 22739-76-0 | C3H8O | 68.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA174850050
Price (MYR)
762.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
762.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsopropanol-d8
Density0.8900g/mL
Deuterium (D)99.0% min.
PackagingGlass Bottle
Physical FormLiquid
View more
For NMR
RUO – Research Use Only