Methyltriethoxysilane, 98+%
Methyltriethoxysilane, 98+%
Methyltriethoxysilane, 98+%
Methyltriethoxysilane, 98+%
Thermo Scientific Chemicals
Methyltriethoxysilane, 98+%
CAS: 2031-67-6 | C7H18O3Si | 178.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA174622500
Price (MYR)
501.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
501.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyltriethoxysilane
Name Note98+%
CAS2031-67-6
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning